Gene Transcripts

Gene Symbol
Rab25
Entrez Gene ID
53868
Gene Name
RAB25, member RAS oncogene family
Gene Aliases
AV012978
Chromosome Location
Chr. 3: 88542028 - 88548279 on Build GRCm38
UniGene ID
Mm.26994
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016899.4 NP_058595.2 4 691
GenBank mRNA AB232620.1 - 4 487
AF119675.1 - 4 627
AK003260.1 - 4 692
BC006624.1 - 4 677 3968810

View Full Product Details