Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006351.3 | NP_006342.2 | 11 | 1283 | |
GenBank mRNA | AF026030.1 | - | 11 | 1100 | |
AF041254.1 | - | 11 | 1141 | ||
AK223062.1 | - | 11 | 1126 | ||
AK289634.1 | - | 11 | 1283 | ||
BC033628.1 | - | 11 | 1113 | 5248163 | |
CR627419.1 | - | 1 | 851 |