Gene Transcripts

Gene Symbol
PFDN6
Entrez Gene ID
10471
Gene Name
prefoldin subunit 6
Gene Aliases
H2-KE2, HKE2, KE-2, PFD6
Chromosome Location
Chr. 6: 33289597 - 33290934 on Build GRCh38
UniGene ID
Hs.446374
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001185181.2 NP_001172110.1 2 330
NM_001265595.1 NP_001252524.1 3 222
NM_001265596.1 NP_001252525.1 3 219
NM_014260.3 NP_055075.1 3 227
GenBank mRNA AK312150.1 - 3 215
BC039033.1 - 2 196 5748799
BC059783.1 - 2 202 5415221
BE794658.1 - 2 183
BM686351.1 - 2 311
BM745352.1 - 3 222
BM792413.1 - 2 204
BM818422.1 - 3 222
BM834357.1 - 3 223
DA256816.1 - 3 215

View Full Product Details