Gene Transcripts
- Gene Symbol
- Cuedc2
- Entrez Gene ID
- 67116
- Gene Name
- CUE domain containing 2
- Gene Aliases
- 3010002G01Rik
- Chromosome Location
- Chr. 19: 46329812 - 46338660 on Build GRCm38
- UniGene ID
- Mm.218848
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164290.1 |
NP_001157762.1
|
8, 9 |
862 |
|
NM_001164291.1 |
NP_001157763.1
|
7, 8 |
754 |
|
NM_001164292.1 |
NP_001157764.1
|
7, 8 |
751 |
|
NM_001164293.1 |
NP_001157765.1
|
7, 8 |
721 |
|
NM_001164294.1 |
NP_001157766.1
|
7, 8 |
700 |
|
NM_001164295.1 |
NP_001157767.1
|
7, 8 |
697 |
|
NM_024192.2 |
NP_077154.2
|
8, 9 |
853 |
|
GenBank mRNA |
AB232359.1 |
- |
6, 7 |
652 |
|
AK003457.1 |
- |
7, 8 |
656 |
|
AK008842.1 |
- |
7, 8 |
717 |
|
AK013884.1 |
- |
7, 8 |
712 |
|
AK183087.1 |
- |
2, 3 |
234 |
|
AK186847.1 |
- |
2, 3 |
236 |
|
BC003442.1 |
- |
8, 9 |
817 |
2650719 |
View Full Product Details