Gene Transcripts
- Gene Symbol
- IFI44
- Entrez Gene ID
- 10561
- Gene Name
- interferon induced protein 44
- Gene Aliases
- MTAP44, TLDC5, p44
- Chromosome Location
- Chr. 1: 78648554 - 78664078 on Build GRCh38
- UniGene ID
- Hs.82316
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006417.4 |
NP_006408.3
|
4, 5 |
808 |
|
NR_135640.1 |
|
3, 4 |
344 |
|
NR_135641.1 |
|
4, 5 |
811 |
|
XM_005270380.3 |
XP_005270437.1
|
4, 5 |
2046 |
|
XM_011540516.2 |
XP_011538818.1
|
4, 5 |
2046 |
|
XM_017000083.1 |
XP_016855572.1
|
4, 5 |
2046 |
|
GenBank mRNA |
AK302527.1 |
- |
4, 5 |
769 |
|
AK316278.1 |
- |
3, 4 |
301 |
|
AK316476.1 |
- |
4, 5 |
766 |
|
AL589497.1 |
- |
3, 4 |
372 |
|
BC022870.1 |
- |
4, 5 |
773 |
4071404 |
View Full Product Details