Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000491.3 | NP_000482.3 | 3 | 347 | |
XM_011542059.2 | XP_011540361.1 | 4 | 498 | ||
GenBank mRNA | BC008983.1 | - | 3 | 289 | |
BQ711602.1 | - | 4 | 509 | ||
X03084.1 | - | 2 | 141 |