Gene Transcripts

Gene Symbol
BTN2A1
Entrez Gene ID
11120
Gene Name
butyrophilin subfamily 2 member A1
Gene Aliases
BK14H9.1, BT2.1, BTF1, BTN2.1, DJ3E1.1
Chromosome Location
Chr. 6: 26457904 - 26476621 on Build GRCh38
UniGene ID
Hs.159028
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001197233.2 NP_001184162.1 4 960
NM_001197234.2 NP_001184163.1 5 1072
NM_007049.4 NP_008980.1 5 1072
NM_078476.3 NP_510961.1 5 1072
GenBank mRNA AB209777.1 - 5 1008
AK297098.1 - 4 914
BC016661.1 - 5 1015 4340392
BX538050.1 - 4 615
U90142.1 - 6 990
U90543.1 - 5 1013

View Full Product Details