Gene Transcripts

Gene Symbol
Ly49i5
Entrez Gene ID
494211
Gene Name
Ly49 inhibitory receptor 5
Gene Aliases
Chromosome Location
Chr. 4: 164345763 - 164357619 on Build Rnor_6.0
UniGene ID
Rn.154805
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009501.2 NP_001009501.1 929
GenBank mRNA AY649836.1 - 929
AY653729.1 - 774
AY846264.1 - 779

View Full Product Details