Gene Transcripts

Gene Symbol
Rspo4
Entrez Gene ID
499918
Gene Name
R-spondin 4
Gene Aliases
RGD1563415
Chromosome Location
Chr. 3: 147358716 - 147391692 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006235262.2 XP_006235324.1 2 400

View Full Product Details