Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199492.1 | NP_001186421.1 | 11 | 1557 | |
NM_014456.4 | NP_055271.2 | 11 | 1599 | ||
NM_145341.3 | NP_663314.1 | 12 | 1683 | ||
GenBank mRNA | AK223623.1 | - | 11 | 1473 | |
AK296753.1 | - | 11 | 1424 | ||
AK312540.1 | - | 11 | 1495 | ||
AK315295.1 | - | 12 | 1561 | ||
BC026104.2 | - | 11 | 1466 | 4798293 | |
BC031049.1 | - | 11 | 1466 | 5277621 | |
BX537500.1 | - | 9 | 5472 | ||
BX647834.1 | - | 11 | 1441 |