Gene Transcripts
- Gene Symbol
- PSMD8
- Entrez Gene ID
- 5714
- Gene Name
- proteasome 26S subunit, non-ATPase 8
- Gene Aliases
- HEL-S-91n, HIP6, HYPF, Nin1p, Rpn12, S14, p31
- Chromosome Location
- Chr. 19: 38374550 - 38383824 on Build GRCh38
- UniGene ID
- Hs.78466
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002812.4 |
NP_002803.2
|
5, 6 |
849 |
|
GenBank mRNA |
AK301771.1 |
- |
5, 6 |
860 |
|
AK304486.1 |
- |
5, 6 |
434 |
|
AK308809.1 |
- |
5, 6 |
938 |
|
BC001164.1 |
- |
5, 6 |
600 |
3528096 |
BC065006.1 |
- |
5, 6 |
588 |
6055235 |
BQ684248.1 |
- |
5, 6 |
612 |
|
BT019688.1 |
- |
5, 6 |
518 |
|
D38047.1 |
- |
5, 6 |
588 |
|
EU668354.1 |
- |
5, 6 |
849 |
|
View Full Product Details