Gene Transcripts

Gene Symbol
ICK
Entrez Gene ID
22858
Gene Name
intestinal cell (MAK-like) kinase
Gene Aliases
ECO, LCK2, MRK
Chromosome Location
Chr. 6: 53001299 - 53061864 on Build GRCh38
UniGene ID
Hs.417022
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014920.3 NP_055735.1 5 721
NM_016513.4 NP_057597.2 6 832
XM_011514419.1 XP_011512721.1 5 721
XM_011514420.2 XP_011512722.1 6 863
XM_011514421.1 XP_011512723.1 6 802
XM_017010485.1 XP_016865974.1 6 1095
XM_017010486.1 XP_016865975.1 7 974
XM_017010487.1 XP_016865976.1 5 640
XM_017010488.1 XP_016865977.1 6 1522
XM_017010489.1 XP_016865978.1 7 1037
XM_017010490.1 XP_016865979.1 6 907
XM_017010491.1 XP_016865980.1 6 868
XM_017010492.1 XP_016865981.1 5 912
GenBank mRNA AB023153.1 - 5 608
AF152469.1 - 5 689
AF225919.1 - 5 551
AK074892.1 - 6 802
BC035807.1 - 5 753
BC136420.1 - 4 382 9020408
BC136421.1 - 4 382 9020409
BC152464.1 - 5 608 8862600

View Full Product Details