Gene Transcripts

Gene Symbol
CNKSR2
Entrez Gene ID
22866
Gene Name
connector enhancer of kinase suppressor of Ras 2
Gene Aliases
CNK2, KSR2, MAGUIN
Chromosome Location
Chr. X: 21374335 - 21654695 on Build GRCh38
UniGene ID
Hs.555917
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168647.1 NP_001162118.1 12 1920
NM_001168648.1 NP_001162119.1 13 2010
NM_001168649.1 NP_001162120.1 12 1863
NM_014927.3 NP_055742.2 13 2010
XM_011545471.2 XP_011543773.1 12 2003
XM_011545472.2 XP_011543774.1 12 1867
XM_017029358.1 XP_016884847.1 11 1777
GenBank mRNA AB020709.1 - 13 1986
AF418269.1 - 13 1566
AF418270.1 - 13 1566
AK056549.1 - 2 362
AK294728.1 - 12 1825
BC126121.1 - 13 1565 8991837
BC136289.1 - 13 1565 9020276
BC143839.1 - 12 1475 9052354

View Full Product Details