Gene Transcripts

Gene Symbol
MB21D2
Entrez Gene ID
151963
Gene Name
Mab-21 domain containing 2
Gene Aliases
C3orf59
Chromosome Location
Chr. 3: 192796815 - 192918161 on Build GRCh38
UniGene ID
Hs.151443
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178496.3 NP_848591.2 2 907
GenBank mRNA AK056276.1 - 1 303
AK290447.1 - 2 602
BC013123.1 - 2 585 4471808
BC036194.1 - 2 891 5267040
BC045582.1 - 2 907 5262015
BX648517.1 - 2 606

View Full Product Details