Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199054.1 | NP_001185983.1 | 6 | 1209 | |
NM_001199055.1 | NP_001185984.1 | 6 | 1060 | ||
NM_001199056.1 | NP_001185985.1 | 6 | 940 | ||
NM_013399.2 | NP_037531.2 | 6 | 1177 | ||
GenBank mRNA | AF131218.1 | - | 5 | 1255 | |
AF195661.1 | - | 6 | 1172 | ||
AK292036.1 | - | 6 | 1135 | ||
AK294257.1 | - | 6 | 1030 | ||
AK302297.1 | - | 6 | 898 | ||
AK308532.1 | - | 4 | 817 | ||
AK311295.1 | - | 2 | 633 | ||
AL136698.1 | - | 6 | 1135 | ||
AL833853.1 | - | 6 | 1209 | ||
BC002882.2 | - | 6 | 1123 | 3941342 | |
BC007604.1 | - | 2 | 711 | ||
DQ167023.1 | - | 6 | 1146 |