Gene Transcripts

Gene Symbol
PRAMEF17
Entrez Gene ID
391004
Gene Name
PRAME family member 17
Gene Aliases
Chromosome Location
Chr. 1: 13389632 - 13392629 on Build GRCh38
UniGene ID
Hs.684629
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099851.3 NP_001093321.1 1 280
GenBank mRNA BC144662.1 - 1 280
BC146858.1 - 1 280 9056786

View Full Product Details