Gene Transcripts

Gene Symbol
NOC3L
Entrez Gene ID
64318
Gene Name
NOC3 like DNA replication regulator
Gene Aliases
AD24, C10orf117, FAD24
Chromosome Location
Chr. 10: 94333226 - 94362976 on Build GRCh38
UniGene ID
Hs.74899
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022451.10 NP_071896.8 3 426
GenBank mRNA AB077992.1 - 3 313
AK091246.1 - 3 383
AK292520.1 - 3 408
BC049850.1 - 3 376 5549517

View Full Product Details