Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145829.1 | NP_001139301.1 | 7 | 358 | |
NM_001145841.1 | NP_001139313.1 | 5 | 319 | ||
NM_003282.3 | NP_003273.1 | 7 | 350 | ||
GenBank mRNA | AW984315.1 | - | 4 | 293 | |
BC032148.2 | - | 7 | 358 | 5020339 | |
BI833431.1 | - | 5 | 328 | ||
BP233165.1 | - | 7 | 372 | ||
BQ926240.1 | - | 7 | 359 | ||
L21715.1 | - | 6 | 323 |