Gene Transcripts

Gene Symbol
PLXND1
Entrez Gene ID
23129
Gene Name
plexin D1
Gene Aliases
PLEXD1
Chromosome Location
Chr. 3: 129555213 - 129606739 on Build GRCh38
UniGene ID
Hs.301685
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015103.2 NP_055918.2 19 3741
XM_011512588.1 XP_011510890.1 19 3656
XM_011512589.1 XP_011510891.1 16 3266
XM_011512590.2 XP_011510892.1 19 3656
XM_011512592.1 XP_011510894.1 7 824
GenBank mRNA AB014520.2 - 19 3820
AY116661.1 - 19 3821
BC150280.1 - 19 3820 9007263

View Full Product Details