Gene Transcripts

Gene Symbol
Mccc1
Entrez Gene ID
72039
Gene Name
methylcrotonoyl-Coenzyme A carboxylase 1 (alpha)
Gene Aliases
1810045E08Rik, 2310058B18Rik, MCCalpha, Mcca, R75106
Chromosome Location
Chr. 3: 35959296 - 36000678 on Build GRCm38
UniGene ID
Mm.249016
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023644.4 NP_076133.3 6 714
GenBank mRNA AF310338.1 - 6 635
AK007782.1 - 6 667
AK031072.1 - 6 675
AK032203.1 - 6 646
AK160184.1 - 4 431
AK168589.1 - 6 603
BC021382.1 - 6 678 5042993

View Full Product Details