Gene Transcripts

Gene Symbol
Dock10
Entrez Gene ID
210293
Gene Name
dedicator of cytokinesis 10
Gene Aliases
9330153B10Rik, A630054M16Rik, Jr4, Jr5, R75174, ZIZ3, Zizimin3, mKIAA0694
Chromosome Location
Chr. 1: 80501068 - 80758704 on Build GRCm38
UniGene ID
Mm.133473
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285927.1 NP_001272856.1 17 2134
NM_175291.4 NP_780500.2 17 2118
GenBank mRNA AK122347.1 - 8 972
AK138722.1 - 17 2120
BC150753.1 - 17 2054 9087664
GU797481.1 - 17 2091

View Full Product Details