Gene Transcripts

Gene Symbol
Lce1h
Entrez Gene ID
67718
Gene Name
late cornified envelope 1H
Gene Aliases
2310066F03Rik, AI426284, Sprrl9
Chromosome Location
Chr. 3: 92763215 - 92765065 on Build GRCm38
UniGene ID
Mm.23784
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026335.2 NP_080611.1 2 561
GenBank mRNA AK009424.1 - 2 561
AK010064.1 - 2 562

View Full Product Details