Gene Transcripts

Gene Symbol
Flnc
Entrez Gene ID
68794
Gene Name
filamin C, gamma
Gene Aliases
1110055E19Rik, ABP-280, ABPL, Fln2
Chromosome Location
Chr. 6: 29433127 - 29461888 on Build GRCm38
UniGene ID
Mm.39046
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081185.1 NP_001074654.1 18 3086
GenBank mRNA BC151097.1 - 18 2891
BC158128.1 - 18 2891 9007396
BC172159.1 - 18 2891

View Full Product Details