Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001013802.3 | NP_001013824.2 | 2 | 327 | |
NM_028387.1 | NP_082663.1 | 2 | 322 | ||
GenBank mRNA | AK004072.1 | - | 2 | 279 | |
AK011824.1 | - | 2 | 317 | ||
AK013491.1 | - | 2 | 324 | ||
AK029184.1 | - | 2 | 302 | ||
AK134694.1 | - | 2 | 329 | ||
AK160433.1 | - | 2 | 302 | ||
AK165589.1 | - | 2 | 317 | ||
BC027552.1 | - | 2 | 162 | ||
BC028443.1 | - | 2 | 162 | ||
BC038317.1 | - | 2 | 248 |