Gene Transcripts
- Gene Symbol
- Slx
- Entrez Gene ID
- 664829
- Gene Name
- Sycp3 like X-linked
- Gene Aliases
- EG664958, Xmr
- Chromosome Location
- Chr. X: 26522657 - 26545565 on Build GRCm38
- UniGene ID
- Mm.489202
- Species
- Mus musculus
- Gene Symbol
- Gm10058
- Entrez Gene ID
- 100039240
- Gene Name
- predicted gene 10058
- Gene Aliases
- ENSMUSG00000059047
- Chromosome Location
- Chr. X: 27924021 - 27946885 on Build GRCm38
- UniGene ID
- Mm.483167
- Species
- Mus musculus
- Gene Symbol
- Gm10096
- Entrez Gene ID
- 100039377
- Gene Name
- predicted gene 10096
- Gene Aliases
- ENSMUSG00000061227, Xmr-like
- Chromosome Location
- Chr. X: 28931807 - 28954702 on Build GRCm38
- Species
- Mus musculus
- Gene Symbol
- Gm10147
- Entrez Gene ID
- 100039324
- Gene Name
- predicted gene 10147
- Gene Aliases
- ENSMUSG00000064334
- Chromosome Location
- Chr. X: 28584995 - 28607874 on Build GRCm38
- Species
- Mus musculus
- Gene Symbol
- Gm10230
- Entrez Gene ID
- 100042175
- Gene Name
- predicted gene 10230
- Gene Aliases
- ENSMUSG00000068103
- Chromosome Location
- Chr. X: 27472033 - 27494914 on Build GRCm38
- UniGene ID
- Mm.483167
- Species
- Mus musculus
- Gene Symbol
- Gm10486
- Entrez Gene ID
- 100039550
- Gene Name
- predicted gene 10486
- Gene Aliases
- ENSMUSG00000073247
- Chromosome Location
- Chr. X: 35067384 - 35090316 on Build GRCm38
- UniGene ID
- Mm.458011
- Species
- Mus musculus
- Gene Symbol
- Gm10487
- Entrez Gene ID
- 100039467
- Gene Name
- predicted gene 10487
- Gene Aliases
- ENSMUSG00000073253
- Chromosome Location
- Chr. X: 30820542 - 30843469 on Build GRCm38
- UniGene ID
- Mm.427330
- Species
- Mus musculus
- Gene Symbol
- Gm10488
- Entrez Gene ID
- 100042109
- Gene Name
- predicted gene 10488
- Gene Aliases
- ENSMUSG00000073257
- Chromosome Location
- Chr. X: 29976314 - 29999162 on Build GRCm38
- UniGene ID
- Mm.432195
- Species
- Mus musculus
- Gene Symbol
- Gm14525
- Entrez Gene ID
- 100039120
- Gene Name
- predicted gene 14525
- Gene Aliases
- OTTMUSG00000017011
- Chromosome Location
- Chr. X: 26672780 - 26702638 on Build GRCm38
- UniGene ID
- Mm.379326
- Species
- Mus musculus
- Gene Symbol
- Gm14632
- Entrez Gene ID
- 100042144
- Gene Name
- predicted gene 14632
- Gene Aliases
- OTTMUSG00000017531
- Chromosome Location
- Chr. X: 30329715 - 30352646 on Build GRCm38
- UniGene ID
- Mm.483167
- Species
- Mus musculus
- Gene Symbol
- Gm14819
- Entrez Gene ID
- 100039585
- Gene Name
- predicted gene 14819
- Gene Aliases
- OTTMUSG00000018086
- Chromosome Location
- Chr. X: 35404215 - 35427094 on Build GRCm38
- UniGene ID
- Mm.483167
- Species
- Mus musculus
- Gene Symbol
- Gm1993
- Entrez Gene ID
- 100038977
- Gene Name
- predicted gene 1993
- Gene Aliases
- Chromosome Location
- Chr. X: 25548501 - 25570449 on Build GRCm38
- Species
- Mus musculus
- Gene Symbol
- Gm2012
- Entrez Gene ID
- 100039030
- Gene Name
- predicted gene 2012
- Gene Aliases
- Chromosome Location
- Chr. X: 26200882 - 26231323 on Build GRCm38
- UniGene ID
- Mm.484125
- Species
- Mus musculus
- Gene Symbol
- Gm2030
- Entrez Gene ID
- 100039065
- Gene Name
- predicted gene 2030
- Gene Aliases
- Chromosome Location
- Chr. X: 26287199 - 26310044 on Build GRCm38
- UniGene ID
- Mm.411645
- Species
- Mus musculus
- Gene Symbol
- Gm4836
- Entrez Gene ID
- 22526
- Gene Name
- predicted gene 4836
- Gene Aliases
- Chromosome Location
- Chr. X: 28270710 - 28293632 on Build GRCm38
- UniGene ID
- Mm.427330
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009529.3 |
NP_033555.1
|
6, 7 |
560 |
|
GenBank mRNA |
BC061169.1 |
- |
6, 7 |
560 |
|
- Gene Symbol
- Gm5168
- Entrez Gene ID
- 382275
- Gene Name
- predicted gene 5168
- Gene Aliases
- EG382275
- Chromosome Location
- Chr. X: 26028214 - 26051041 on Build GRCm38
- UniGene ID
- Mm.370361
- Species
- Mus musculus
- Gene Symbol
- Gm5169
- Entrez Gene ID
- 382277
- Gene Name
- predicted gene 5169
- Gene Aliases
- Xmr
- Chromosome Location
- Chr. X: 25277487 - 25301455 on Build GRCm38
- UniGene ID
- Mm.379326
- Species
- Mus musculus
- Gene Symbol
- Gm5934
- Entrez Gene ID
- 546272
- Gene Name
- predicted gene 5934
- Gene Aliases
- EG546272
- Chromosome Location
- Chr. X: 24474308 - 24499163 on Build GRCm38
- Species
- Mus musculus
- Gene Symbol
- Gm5935
- Entrez Gene ID
- 546282
- Gene Name
- predicted gene 5935
- Gene Aliases
- EG546282
- Chromosome Location
- Chr. X: 24753162 - 24775164 on Build GRCm38
- UniGene ID
- Mm.391219
- Species
- Mus musculus
- Gene Symbol
- Gm6121
- Entrez Gene ID
- 619991
- Gene Name
- predicted gene 6121
- Gene Aliases
- Chromosome Location
- Chr. X: 26912743 - 26935669 on Build GRCm38
- UniGene ID
- Mm.483167
- Species
- Mus musculus
View Full Product Details