Gene Transcripts

Gene Symbol
B3gnt3
Entrez Gene ID
290638
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 3
Gene Aliases
Chromosome Location
Chr. 16: 20123948 - 20133322 on Build Rnor_6.0
UniGene ID
Rn.218499
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106068.1 NP_001099538.1 3 808
XM_008771081.1 XP_008769303.1 2 758

View Full Product Details