Gene Transcripts
- Gene Symbol
- ZFYVE26
- Entrez Gene ID
- 23503
- Gene Name
- zinc finger FYVE-type containing 26
- Gene Aliases
- FYVE-CENT, SPG15
- Chromosome Location
- Chr. 14: 67728892 - 67817293 on Build GRCh38
- UniGene ID
- Hs.98041
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015346.3 |
NP_056161.2
|
34, 35 |
6512 |
|
XM_017021124.1 |
XP_016876613.1
|
34, 35 |
6531 |
|
XM_017021125.1 |
XP_016876614.1
|
34, 35 |
6531 |
|
XM_017021126.1 |
XP_016876615.1
|
33, 34 |
7145 |
|
XM_017021127.1 |
XP_016876616.1
|
23, 24 |
4227 |
|
XM_017021128.1 |
XP_016876617.1
|
23, 24 |
4080 |
|
GenBank mRNA |
AB002319.1 |
- |
19, 20 |
3382 |
|
AB425197.1 |
- |
34, 35 |
6500 |
|
AK024743.1 |
- |
6, 7 |
874 |
|
BC033235.1 |
- |
1, 2 |
207 |
|
BC144594.1 |
- |
32, 33 |
5816 |
|
BX537886.1 |
- |
34, 35 |
6506 |
|
BX538025.1 |
- |
30, 31 |
6005 |
|
BX648683.1 |
- |
34, 35 |
6510 |
|
CR749276.1 |
- |
35, 36 |
6447 |
|
View Full Product Details