Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001756.3 | NP_001747.2 | 4 | 1097 | |
GenBank mRNA | AK225591.1 | - | 5 | 1007 | |
AK290821.1 | - | 4 | 1082 | ||
AK313760.1 | - | 4 | 1082 | ||
BC017462.1 | - | 4 | 1051 | ||
BC056259.1 | - | 4 | 1072 | 5810260 | |
BC058021.1 | - | 4 | 1081 | 6714173 | |
J02943.1 | - | 3 | 1028 |