Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252156.1 | NP_001239085.1 | 16 | 2343 | |
NM_015465.4 | NP_056280.2 | 16 | 2346 | ||
GenBank mRNA | AK074066.2 | - | 16 | 2318 | |
AK315101.1 | - | 16 | 2331 | ||
AL117665.1 | - | 11 | 1408 | ||
AY063750.1 | - | 16 | 2263 | ||
BC008776.2 | - | 4 | 504 | ||
BC014147.2 | - | 4 | 504 | ||
BC113614.1 | - | 16 | 2302 | 8322666 | |
BC143724.1 | - | 16 | 2299 | 9052238 |