Gene Transcripts

Gene Symbol
DKKL1
Entrez Gene ID
27120
Gene Name
dickkopf like acrosomal protein 1
Gene Aliases
CT34, SGY, SGY-1, SGY1
Chromosome Location
Chr. 19: 49360520 - 49375132 on Build GRCh38
UniGene ID
Hs.515855
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001197302.1 NP_001184231.1 4 581
NM_014419.3 NP_055234.1 4 620
XM_006723142.2 XP_006723205.1 4 466
XM_011526724.1 XP_011525026.1 4 490
XM_011526725.1 XP_011525027.1 4 440
XM_011526726.2 XP_011525028.1 4 700
XM_011526729.1 XP_011525031.1 4 461
XM_017026593.1 XP_016882082.1 4 473
GenBank mRNA AF177398.1 - 4 471
AY358922.1 - 4 620
BC030581.2 - 4 500 4836991
BI826695.1 - 4 581

View Full Product Details