Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014438.4 | NP_055253.2 | 3 | 182 | |
NM_173178.2 | NP_775270.1 | 3 | 182 | ||
XM_011510962.1 | XP_011509264.1 | 3 | 331 | ||
GenBank mRNA | AF200494.1 | - | 3 | 185 | |
AF201833.1 | - | 3 | 185 | ||
BC101831.1 | - | 2 | 136 | 8069337 | |
BC101833.1 | - | 2 | 136 | 8069339 |