Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014448.3 | NP_055263.2 | 12 | 1887 | |
XM_017001049.1 | XP_016856538.1 | 12 | 2488 | ||
XM_017001050.1 | XP_016856539.1 | 12 | 1693 | ||
XM_017001051.1 | XP_016856540.1 | 12 | 1898 | ||
GenBank mRNA | AK091910.1 | - | 5 | 1323 | |
AK095984.1 | - | 12 | 1794 | ||
AK296150.1 | - | 7 | 785 | ||
BC002681.1 | - | 11 | 1174 | 3613623 | |
BC051838.1 | - | 10 | 1194 | 6191377 | |
BT007270.1 | - | 7 | 775 | ||
D89016.1 | - | 9 | 1202 |