Gene Transcripts

Gene Symbol
C19orf53
Entrez Gene ID
28974
Gene Name
chromosome 19 open reading frame 53
Gene Aliases
HSPC023, LYDG10
Chromosome Location
Chr. 19: 13774443 - 13778773 on Build GRCh38
UniGene ID
Hs.231616
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014047.2 NP_054766.1 1, 2 141
GenBank mRNA AF078852.1 - 1, 2 115
AF086483.1 - 1, 2 113
AK311855.1 - 1, 2 106
BC015465.2 - 1, 2 116 3914049
DA768829.1 - 1, 2 142

View Full Product Details