Gene Transcripts

Gene Symbol
CD70
Entrez Gene ID
970
Gene Name
CD70 molecule
Gene Aliases
CD27L, CD27LG, TNFSF7, TNLG8A
Chromosome Location
Chr. 19: 6581646 - 6591152 on Build GRCh38
UniGene ID
Hs.501497
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252.4 NP_001243.1 3 685
GenBank mRNA BC000725.2 - 3 631
BT007211.1 - 3 535
L08096.1 - 3 685
S69339.1 - 3 672

View Full Product Details