Gene Transcripts
- Gene Symbol
- RAPGEFL1
- Entrez Gene ID
- 51195
- Gene Name
- Rap guanine nucleotide exchange factor like 1
- Gene Aliases
- Link-GEFII
- Chromosome Location
- Chr. 17: 40177010 - 40195656 on Build GRCh38
- UniGene ID
- Hs.632254
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001303533.1 |
NP_001290462.1
|
3, 4 |
337 |
|
NM_001303534.1 |
NP_001290463.1
|
3, 4 |
432 |
|
NM_016339.4 |
NP_057423.1
|
3, 4 |
617 |
|
XM_006721932.3 |
XP_006721995.1
|
3, 4 |
1111 |
|
XM_006721935.3 |
XP_006721998.1
|
4, 5 |
388 |
|
XM_006721937.3 |
XP_006722000.1
|
3, 4 |
2213 |
|
GenBank mRNA |
AF117946.1 |
- |
3, 4 |
252 |
|
AK304160.1 |
- |
3, 4 |
432 |
|
AK313492.1 |
- |
3, 4 |
292 |
|
BC029956.1 |
- |
3, 4 |
1111 |
|
BC115374.1 |
- |
1, 2 |
146 |
40028278 |
BC115375.1 |
- |
1, 2 |
146 |
40028280 |
DB265550.1 |
- |
3, 4 |
339 |
|
View Full Product Details