Gene Transcripts
- Gene Symbol
- RAB4B
- Entrez Gene ID
- 53916
- Gene Name
- RAB4B, member RAS oncogene family
- Gene Aliases
- Chromosome Location
- Chr. 19: 40778219 - 40796944 on Build GRCh38
- UniGene ID
- Hs.631539
- Species
- Homo sapiens
- Gene Symbol
- MIA-RAB4B
- Entrez Gene ID
- 100529262
- Gene Name
- MIA-RAB4B readthrough (NMD candidate)
- Gene Aliases
- Chromosome Location
- Chr. 19: 40775537 - 40796944 on Build GRCh38
- UniGene ID
- Hs.631539
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037775.1 |
|
5 |
531 |
|
GenBank mRNA |
AF087861.1 |
- |
4 |
312 |
|
AK302147.1 |
- |
6 |
947 |
|
- Gene Symbol
- RAB4B-EGLN2
- Entrez Gene ID
- 100529264
- Gene Name
- RAB4B-EGLN2 readthrough (NMD candidate)
- Gene Aliases
- RERT-lncRNA
- Chromosome Location
- Chr. 19: 40778219 - 40808441 on Build GRCh38
- UniGene ID
- Hs.631539
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037791.1 |
|
3 |
326 |
|
GenBank mRNA |
AK291385.1 |
- |
3 |
274 |
|
View Full Product Details