Gene Transcripts
- Gene Symbol
- ZDHHC13
- Entrez Gene ID
- 54503
- Gene Name
- zinc finger DHHC-type containing 13
- Gene Aliases
- HIP14L, HIP3RP
- Chromosome Location
- Chr. 11: 19117099 - 19176420 on Build GRCh38
- UniGene ID
- Hs.188569
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001001483.2 |
NP_001001483.1
|
8, 9 |
971 |
|
NM_019028.2 |
NP_061901.2
|
9, 10 |
1117 |
|
XM_005252996.2 |
XP_005253053.1
|
9, 10 |
1089 |
|
XM_011520193.1 |
XP_011518495.1
|
7, 8 |
814 |
|
XM_011520194.2 |
XP_011518496.1
|
9, 10 |
1092 |
|
XM_011520195.1 |
XP_011518497.1
|
9, 10 |
1027 |
|
GenBank mRNA |
AB024495.1 |
- |
9, 10 |
1065 |
|
AB097014.1 |
- |
9, 10 |
1115 |
|
AB097027.1 |
- |
9, 10 |
1115 |
|
AK001714.1 |
- |
9, 10 |
1115 |
|
AK001831.1 |
- |
4, 5 |
489 |
|
AK291348.1 |
- |
9, 10 |
1092 |
|
AL833714.1 |
- |
5, 6 |
567 |
|
BC036020.1 |
- |
9, 10 |
1102 |
5272445 |
BC050690.1 |
- |
8, 9 |
971 |
6171322 |
BC056152.1 |
- |
9, 10 |
1117 |
6170701 |
BX648156.1 |
- |
8, 9 |
961 |
|
View Full Product Details