Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017639.3 | NP_060109.2 | 22 | 5618 | |
XM_011532045.1 | XP_011530347.1 | 17 | 7406 | ||
XM_011532046.2 | XP_011530348.1 | 16 | 5062 | ||
XM_011532049.2 | XP_011530351.1 | 9 | 2912 | ||
GenBank mRNA | AK095103.1 | - | 5 | 955 | |
AY354497.1 | - | 22 | 5618 | ||
BX647835.1 | - | 5 | 594 |