Gene Transcripts

Gene Symbol
PIGG
Entrez Gene ID
54872
Gene Name
phosphatidylinositol glycan anchor biosynthesis class G
Gene Aliases
GPI7, LAS21, MRT53, PRO4405, RLGS1930
Chromosome Location
Chr. 4: 499200 - 539921 on Build GRCh38
UniGene ID
Hs.7099
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127178.2 NP_001120650.1 5 1006
NM_001289051.1 NP_001275980.1 5 815
NM_001289052.1 NP_001275981.1 3 607
NM_001289053.1 NP_001275982.1 5 1564
NM_001289055.1 NP_001275984.1 6 1076
NM_001289057.1 NP_001275986.1 5 1242
NM_017733.4 NP_060203.3 5 1006
NR_110293.1 5 1006
XM_005272284.2 XP_005272341.1 5 1240
XM_011513490.2 XP_011511792.1 5 969
XM_011513491.1 XP_011511793.1 6 1036
XM_011513493.1 XP_011511795.1 5 1006
XM_011513494.2 XP_011511796.1 5 1006
XM_017008321.1 XP_016863810.1 5 1240
XM_017008323.1 XP_016863812.1 5 1240
XM_017008324.1 XP_016863813.1 5 1240
XM_017008325.1 XP_016863814.1 5 953
XM_017008326.1 XP_016863815.1 5 1006
GenBank mRNA AB162713.1 - 5 976
AK074715.1 - 5 778
AK074815.1 - 5 985
AK097244.1 - 5 1203
AK296507.1 - 6 1067
AY358538.1 - 5 948
BC050262.1 - 5 1476
BC110878.1 - 3 549 5750347
BC144576.1 - 5 963

View Full Product Details