Gene Transcripts
- Gene Symbol
- TTC19
- Entrez Gene ID
- 54902
- Gene Name
- tetratricopeptide repeat domain 19
- Gene Aliases
- 2010204O13Rik, MC3DN2
- Chromosome Location
- Chr. 17: 15999380 - 16045428 on Build GRCh38
- UniGene ID
- Hs.462316
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271420.1 |
NP_001258349.1
|
5, 6 |
1130 |
|
NM_017775.3 |
NP_060245.3
|
5, 6 |
993 |
|
XM_017024801.1 |
XP_016880290.1
|
5, 6 |
906 |
|
XM_017024802.1 |
XP_016880291.1
|
5, 6 |
906 |
|
GenBank mRNA |
AK000350.1 |
- |
3, 4 |
209 |
|
AK055780.1 |
- |
5, 6 |
686 |
|
AK056878.1 |
- |
5, 6 |
905 |
|
AK297783.1 |
- |
5, 6 |
549 |
|
BC011698.2 |
- |
5, 6 |
532 |
|
BC073796.1 |
- |
5, 6 |
536 |
|
BC105128.1 |
- |
5, 6 |
993 |
8144131 |
BC112107.1 |
- |
5, 6 |
993 |
8327575 |
View Full Product Details