Gene Transcripts

Gene Symbol
PLXNA3
Entrez Gene ID
55558
Gene Name
plexin A3
Gene Aliases
6.3, HSSEXGENE, PLXN3, PLXN4, XAP-6
Chromosome Location
Chr. X: 154458281 - 154473646 on Build GRCh38
UniGene ID
Hs.632839
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017514.4 NP_059984.3 17, 18 3282
XM_005274705.4 XP_005274762.2 17, 18 3222
XM_005274706.4 XP_005274763.2 17, 18 2954
XM_006724829.3 XP_006724892.1 17, 18 3186
XM_011531183.2 XP_011529485.1 16, 17 2646
GenBank mRNA BQ718472.1 - 4, 5 477
X87852.1 - 17, 18 3291

View Full Product Details