Gene Transcripts

Gene Symbol
LUC7L
Entrez Gene ID
55692
Gene Name
LUC7 like
Gene Aliases
LUC7B1, Luc7, SR+89, hLuc7B1
Chromosome Location
Chr. 16: 188969 - 229482 on Build GRCh38
UniGene ID
Hs.16803
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320226.1 NP_001307155.1 4 498
NM_018032.4 NP_060502.1 4 498
NM_201412.2 NP_958815.1 4 498
XM_005255427.3 XP_005255484.1 5 535
XM_005255429.3 XP_005255486.1 6 581
XM_011522561.2 XP_011520863.1 3 337
XM_017023437.1 XP_016878926.1 4 1561
XM_017023438.1 XP_016878927.1 5 1625
XM_017023439.1 XP_016878928.1 4 1561
XM_017023440.1 XP_016878929.1 5 1625
GenBank mRNA AJ404326.1 - 4 426
AK001093.1 - 4 426
AK097974.1 - 2 1276
AK223084.1 - 4 429
AY005111.1 - 4 466
BC003194.1 - 4 443 3536109
BC065198.1 - 4 1715 5212471
BX647161.1 - 4 1496

View Full Product Details