Gene Transcripts

Gene Symbol
DNAJC11
Entrez Gene ID
55735
Gene Name
DnaJ heat shock protein family (Hsp40) member C11
Gene Aliases
dJ126A5.1
Chromosome Location
Chr. 1: 6634168 - 6701906 on Build GRCh38
UniGene ID
Hs.462640
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018198.3 NP_060668.2 13 1453
XM_017001745.1 XP_016857234.1 12 1306
GenBank mRNA AF306695.1 - 13 1367
AK001599.1 - 13 1362
AK025491.1 - 3 127
AK093709.1 - 1 608
AK095386.1 - 4 1175
AK294539.1 - 12 1276
AK298378.1 - 13 1339
AL833841.1 - 13 1327
AY336749.1 - 13 1363
BC006086.1 - 13 1349 3946414
BC008772.1 - 13 1354
BC014145.2 - 12 1205 4762069

View Full Product Details