Gene Transcripts

Gene Symbol
BCAP29
Entrez Gene ID
55973
Gene Name
B-cell receptor-associated protein 29
Gene Aliases
BAP29
Chromosome Location
Chr. 7: 107579977 - 107623317 on Build GRCh38
UniGene ID
Hs.303787
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008405.2 NP_001008405.1 3 311
NM_018844.3 NP_061332.2 4 636
NR_027830.1 3 530
XM_006716051.2 XP_006716114.1 4 632
XM_011516406.2 XP_011514708.1 4 395
XM_011516407.1 XP_011514709.1 4 632
XM_011516408.2 XP_011514710.1 4 632
XM_011516409.2 XP_011514711.1 4 632
XM_017012436.1 XP_016867925.1 4 632
XM_017012437.1 XP_016867926.1 4 632
XM_017012438.1 XP_016867927.1 4 632
GenBank mRNA AF126020.1 - 4 343
AK289401.1 - 4 393
AK294824.1 - 3 261
AK298594.1 - 1 37
BC008478.1 - 4 348 4294273
BM023571.1 - 1 106
BT006981.1 - 3 297
BX647653.1 - 4 347
CK820004.1 - 2 168

View Full Product Details