Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001038707.1 | NP_001033796.1 | 4 | 648 | |
NM_020239.3 | NP_064624.1 | 3 | 512 | ||
XM_017001847.1 | XP_016857336.1 | 4 | 592 | ||
GenBank mRNA | AF187845.2 | - | 3 | 437 | |
AF286041.1 | - | 2 | 72 | ||
AF286592.1 | - | 2 | 989 | ||
BC012796.2 | - | 2 | 256 | 3924598 | |
BC041604.1 | - | 3 | 474 | 5473010 | |
BP340339.1 | - | 3 | 513 | ||
BX339337.2 | - | 4 | 561 |