Gene Transcripts
- Gene Symbol
- PDZD4
- Entrez Gene ID
- 57595
- Gene Name
- PDZ domain containing 4
- Gene Aliases
- LNX5, LU1, PDZK4, PDZRN4L
- Chromosome Location
- Chr. X: 153802166 - 153830567 on Build GRCh38
- UniGene ID
- Hs.92732
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001303512.1 |
NP_001290441.1
|
5, 6 |
835 |
|
NM_001303513.1 |
NP_001290442.1
|
6, 7 |
1078 |
|
NM_001303514.1 |
NP_001290443.1
|
3, 4 |
490 |
|
NM_001303515.1 |
NP_001290444.1
|
5, 6 |
670 |
|
NM_001303516.1 |
NP_001290445.1
|
5, 6 |
652 |
|
NM_032512.3 |
NP_115901.2
|
5, 6 |
817 |
|
GenBank mRNA |
AB040877.1 |
- |
2, 3 |
155 |
|
AB103478.1 |
- |
5, 6 |
798 |
|
AK091444.1 |
- |
5, 6 |
543 |
|
AK123229.1 |
- |
5, 6 |
572 |
|
AK123822.1 |
- |
5, 6 |
670 |
|
AK127016.1 |
- |
5, 6 |
817 |
|
AK309212.1 |
- |
6, 7 |
883 |
|
BC020347.1 |
- |
4, 5 |
380 |
|
BC117277.1 |
- |
5, 6 |
627 |
40125828 |
BC143459.1 |
- |
3, 4 |
281 |
9051967 |
BC143545.1 |
- |
5, 6 |
626 |
9052056 |
View Full Product Details