Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020961.3 | NP_066012.1 | 11 | 1322 | |
XM_005263166.4 | XP_005263223.1 | 10 | 1106 | ||
XM_011532162.2 | XP_011530464.1 | 11 | 1156 | ||
GenBank mRNA | AB046847.1 | - | 11 | 1277 | |
AK055555.1 | - | 11 | 1300 | ||
AK296056.1 | - | 9 | 942 | ||
BC006565.2 | - | 11 | 1273 | 3010654 | |
BC007449.2 | - | 11 | 1273 | 3010654 |