Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022773.2 | NP_073610.2 | 8 | 1181 | |
NR_033645.1 | 9 | 1310 | |||
NR_033646.1 | 10 | 1488 | |||
XM_006720928.2 | XP_006720991.1 | 8 | 1454 | ||
XM_006720929.3 | XP_006720992.1 | 9 | 1335 | ||
XM_011522614.2 | XP_011520916.1 | 8 | 1179 | ||
XM_011522617.2 | XP_011520919.1 | 6 | 830 | ||
XM_017023575.1 | XP_016879064.1 | 8 | 1179 | ||
XM_017023576.1 | XP_016879065.1 | 7 | 945 | ||
XM_017023577.1 | XP_016879066.1 | 9 | 1610 | ||
XM_017023578.1 | XP_016879067.1 | 7 | 1030 | ||
XM_017023579.1 | XP_016879068.1 | 7 | 1038 | ||
XM_017023580.1 | XP_016879069.1 | 10 | 1760 | ||
GenBank mRNA | AB075873.1 | - | 8 | 1165 | |
AK022743.1 | - | 8 | 1179 | ||
AK025955.1 | - | 8 | 1113 | ||
AK093050.1 | - | 5 | 688 | ||
AK125201.1 | - | 9 | 1312 | ||
AK225065.1 | - | 8 | 1179 | ||
AK225406.1 | - | 8 | 1166 | ||
AL832177.1 | - | 2 | 2799 | ||
BC010738.1 | - | 7 | 946 |