Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022835.2 | NP_073746.2 | 19 | 3986 | |
XM_005259163.2 | XP_005259220.1 | 19 | 3807 | ||
XM_006723334.2 | XP_006723397.1 | 19 | 4337 | ||
XM_011527232.2 | XP_011525534.1 | 19 | 3913 | ||
XM_017027150.1 | XP_016882639.1 | 18 | 4478 | ||
XM_017027151.1 | XP_016882640.1 | 18 | 4469 | ||
GenBank mRNA | AK024429.1 | - | 19 | 3986 | |
AK302476.1 | - | 19 | 3610 | ||
AL832850.1 | - | 1 | 577 | ||
BC013426.1 | - | 9 | 2477 | ||
BC015174.1 | - | 1 | 86 | ||
BC075858.1 | - | 15 | 3264 |