Gene Transcripts
- Gene Symbol
- CSF2
- Entrez Gene ID
- 1437
- Gene Name
- colony stimulating factor 2
- Gene Aliases
- GMCSF
- Chromosome Location
- Chr. 5: 132073792 - 132076170 on Build GRCh38
- UniGene ID
- Hs.1349
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000758.3 |
NP_000749.2
|
1, 2 |
195 |
|
GenBank mRNA |
AF510855.1 |
- |
1, 2 |
112 |
|
AY720441.1 |
- |
1, 2 |
172 |
|
BC108724.1 |
- |
1, 2 |
182 |
3886571 |
BC113924.1 |
- |
1, 2 |
186 |
40084800 |
BC113999.1 |
- |
1, 2 |
186 |
40084798 |
KP271009.1 |
- |
1, 2 |
112 |
|
M10663.1 |
- |
1, 2 |
171 |
|
M11220.1 |
- |
1, 2 |
195 |
|
M11734.1 |
- |
1, 2 |
178 |
|
View Full Product Details